二苯基乙酰氯 ≥98.0% (by GC, titration analysis)

供应商: TCI
Danger

Synonyms: 2,2-Diphenylacetyl chloride

D2763-25G D2763-5G
TCIAD2763-25GEA 0 CNY
TCIAD2763-25G TCIAD2763-5G
二苯基乙酰氯 ≥98.0% (by GC, titration analysis)
二苯基乙酰氯
分子式: C₁₄H₁₁ClO
MDL编号: MFCD00013655
CAS号: 1871-76-7
EINECS: 217-493-5
UN号: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc748fc52b5613859ebd7d645340cf29fa32fcc2&vid=dc748fc52b5613859ebd7d645340cf29fa32fcc2&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 51.0 to 55.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR