(-)-O,O′-二苯甲酰-L-酒石酸 ≥98.0% (by titrimetric analysis)
供应商: TCI
|
Warning
|
Synonyms:
(2R,3R)-(-)-Di-O-benzoyl-tartaric acid monohydrate
D1354-25G
D1354-500G
TCIAD1354-25GEA
0
CNY
TCIAD1354-25G
TCIAD1354-500G
(-)-O,O′-二苯甲酰-L-酒石酸 ≥98.0% (by titrimetric analysis)
(-)-O,O′-二苯甲酰-L-酒石酸
分子式:
C₁₈H₁₄O₈·1H₂O 分子量: 376.33 g/mol (358.31 anhy) |
MDL编号:
MFCD00149119 CAS号: 62708-56-9 EINECS: 220-374-0 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % (calcd.on anh.substance) |
Specific rotation [a]20/D | -106.0 to -111.0 deg (C=1, EtOH) |
Water | 4.7 to 5.1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...