2′-脱氧胞苷盐酸盐 ≥98.0% (by titrimetric analysis)

供应商: TCI

Synonyms: 1-(2-脱氧-β-D-呋喃核糖基)胞嘧啶, 2'-脱氧胞嘧啶核苷

D0048-25G D0048-5G
TCIAD0048-25GEA 0 CNY
TCIAD0048-25G TCIAD0048-5G
2′-脱氧胞苷盐酸盐 ≥98.0% (by titrimetric analysis)
2′-脱氧胞苷盐酸盐
分子式: C₉H₁₄ClN₃O₄
存储条件: Ambient
MDL编号: MFCD00012840
CAS号: 3992-42-5
EINECS: 223-639-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20d725fcbb5942779ab7351354b268290355867d&vid=20d725fcbb5942779ab7351354b268290355867d&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Specific rotation [a]20/D +55.0 to +59.0 deg (C=1, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR