6-Azauracil ≥99.0% (通过HPLC,滴定分析)
供应商: TCI EUROPE
Synonyms:
6-Aza-2,4-dihydroxypyrimidine
, 1,2,4-Triazine-3,5(2H,4H)-dione
, 3,5-Dihydroxy-1,2,4-triazine
TCIAA0558-1GEA
0
CNY
TCIAA0558-1G
TCIAA0558-5G
6-Azauracil ≥99.0% (通过HPLC,滴定分析)
6-Azauracil
分子式:
C₃H₃N₃O₂ 分子量: 113,08 g/mol |
MDL编号:
MFCD00006456 CAS号: 461-89-2 EINECS: 207-318-0 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Purity (Neutralization titration) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...