6-Methoxypurine ≥98.0% (by HPLC, titration analysis)

供应商: TCI

Synonyms: 6-Methoxy-9H-purine

M1925-1G M1925-5G
TCIAM1925-1GEA 0 CNY
TCIAM1925-1G TCIAM1925-5G
6-Methoxypurine ≥98.0% (by HPLC, titration analysis)
6-Methoxypurine
分子式: C₆H₆N₄O
分子量: 150.1400 g/mol
溶点: 196°C
MDL编号: MFCD00005574
CAS号: 1074-89-1
EINECS: 214-047-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f95836e28aa61b8ae89e7eba0714fdf5ce6e8c45&vid=f95836e28aa61b8ae89e7eba0714fdf5ce6e8c45&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR