2-Amino-6-iodopurine ≥98.0% (by HPLC, titration analysis)

供应商: TCI

Synonyms: 6-Iodo-9H-purin-2-amine

A2068-25G A2068-5G
TCIAA2068-25GEA 0 CNY
TCIAA2068-25G TCIAA2068-5G
2-Amino-6-iodopurine ≥98.0% (by HPLC, titration analysis)
2-Amino-6-iodopurine
分子式: C₅H₄IN₅
溶点: 240°C
MDL编号: MFCD08706375
CAS号: 19690-23-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2081f8fe26eb603b752ff710d39ee4cba2f66e6&vid=f2081f8fe26eb603b752ff710d39ee4cba2f66e6&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystalline
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR