Cresol red sodium salt
供应商: TCI
Synonyms:
Cresol Red sodium, Sodium 4-(3-(4-hydroxy-3-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-2-methylphenolate
S0046-1G
S0046-25G
TCIAS0046-1GEA
0
CNY
TCIAS0046-1G
TCIAS0046-25G
Cresol red sodium salt
Cresol red sodium salt
分子式:
C₂₁H₁₇NaO₅S 分子量: 404.42 g/mol 存储条件: Ambient |
MDL编号:
MFCD00001618 CAS号: 62625-29-0 EINECS: 263-654-8 |
|
产品技术规格
Appearance | Dark red to Brown powder to crystal |
Transition interval (pH) | (yellow)7.2 to 8.8 (red) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...