D-2-脱氧葡萄糖 ≥97.0% (by GC)

供应商: TCI

Synonyms: 2-脱氧-D-葡糖

D0051-1G D0051-5G
TCIAD0051-1GEA 0 CNY
TCIAD0051-1G TCIAD0051-5G
D-2-脱氧葡萄糖 ≥97.0% (by GC)
D-2-脱氧葡萄糖
分子式: C₆H₁₂O₅
分子量: 164.16 g/mol
MDL编号: MFCD00151328
CAS号: 154-17-6
EINECS: 205-823-0
Merck Index: 14,02904
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87caa302445f6f20523c3de710090f7bc9f797cf&vid=87caa302445f6f20523c3de710090f7bc9f797cf&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 % (total of isomers )
Specific rotation [a]20/D +44.0 to +48.0 deg (C=1, H2O)
Solubility in Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR