双十八烷基二甲基溴化铵 ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 二甲基二(十八烷基)溴化铵

D1975-25G D1975-250G
TCIAD1975-25GEA 0 CNY
TCIAD1975-25G TCIAD1975-250G
双十八烷基二甲基溴化铵 ≥98.0% (by titrimetric analysis)
双十八烷基二甲基溴化铵
分子式: C₃₈H₈₀BrN
分子量: 630.952 g/mol
MDL编号: MFCD00041975
CAS号: 3700-67-2
EINECS: 223-037-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2316d3d3311ee05a69d1dfeb111f4caf7b2162e&vid=b2316d3d3311ee05a69d1dfeb111f4caf7b2162e&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 154.0 to 162.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR