5-Iodo-2'-deoxyuridine (Idoxuridine) ≥99.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-iodopyrimidine-2,4(1H,3H)-dione, Idoxuridine, (+)-5-Iodo-2'-deoxyuridine

I0258-1G I0258-25G I0258-5G
TCIAI0258-1GEA 0 CNY
TCIAI0258-1G TCIAI0258-25G TCIAI0258-5G
5-Iodo-2'-deoxyuridine (Idoxuridine) ≥99.0% (by titrimetric analysis)
5-Iodo-2'-deoxyuridine (Idoxuridine)
分子式: C₉H₁₁IN₂O₅
分子量: 354.10 g/mol
存储条件: Ambient
MDL编号: MFCD00134656
CAS号: 54-42-2
EINECS: 200-207-8
Merck Index: 14,04891
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbf2b32a6a393135c6d12376b73b9ebb3a18a062&vid=bbf2b32a6a393135c6d12376b73b9ebb3a18a062&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 99.0 %
Specific rotation [a]20/D +28.0 to +31.0 deg (C=1, 1mol/L NaOH sol.)
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR