尿嘧啶核苷 ≥98.0% (by HPLC, titration analysis)

供应商: TCI

Synonyms: 尿嘧啶-1-β-D-呋喃核糖苷, 尿苷

U0020-25G U0020-5G
TCIAU0020-25GEA 0 CNY
TCIAU0020-25G TCIAU0020-5G
尿嘧啶核苷 ≥98.0% (by HPLC, titration analysis)
尿嘧啶核苷
分子式: C₉H₁₂N₂O₆
分子量: 244.20 g/mol
存储条件: Ambient
MDL编号: MFCD00006526
CAS号: 58-96-8
EINECS: 200-407-5
Merck Index: 14,09877
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a324c3416862b10a8557346e3935bd0e11b0e4f9&vid=a324c3416862b10a8557346e3935bd0e11b0e4f9&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 165.0 to 169.0 deg-C
Specific rotation [a]20/D +8.0 to +10.0 deg (C=2, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR