(±)-薄荷醇 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 薄荷醇, 2-异丙基-5-甲基环己醇, 六氢百里香酚

M0321-25ML M0321-500ML
TCIAM0321-25MLEA 0 CNY
TCIAM0321-25ML TCIAM0321-500ML
(±)-薄荷醇 ≥98.0% (by GC)
(±)-薄荷醇
分子式: C₁₀H₂₀O
分子量: 156.27 g/mol
存储条件: Ambient
MDL编号: MFCD00001484
CAS号: 89-78-1
EINECS: 201-939-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91fd81a8d5b9f34395e220d5a3b1244ff9f102f9&vid=91fd81a8d5b9f34395e220d5a3b1244ff9f102f9&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR