磺乙酰胺 ≥98.0% (by HPLC, titration analysis)

供应商: TCI

Synonyms: N1-乙酰磺胺, N-(4-Aminophenylsulfonyl)acetamide, N-(4-氨基苯磺酰)乙酰胺

S0577-25G S0577-250G
TCIAS0577-25GEA 0 CNY
TCIAS0577-25G TCIAS0577-250G
磺乙酰胺 ≥98.0% (by HPLC, titration analysis)
磺乙酰胺
分子式: C₈H₁₀N₂O₃S
分子量: 214.24 g/mol
MDL编号: MFCD00066501
CAS号: 144-80-9
EINECS: 205-640-6
Merck Index: 14,08899
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d65169aa764ef06070e91d926d569269e18d32ed&vid=d65169aa764ef06070e91d926d569269e18d32ed&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 179.0 to 184.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR