3,5-Dibromo-2-methylthiophene ≥95.0% (by GC)
供应商: TCI
D3999-25G
D3999-5G
TCIAD3999-25GEA
0
CNY
TCIAD3999-25G
TCIAD3999-5G
3,5-Dibromo-2-methylthiophene ≥95.0% (by GC)
3,5-Dibromo-2-methylthiophene
分子式:
C₅H₄Br₂S 分子量: 255.9583 g/mol 沸点: 230°C 溶点: –15°C 存储条件: Refrigerator |
MDL编号:
MFCD07781197 CAS号: 29421-73-6 EINECS: 249-617-9 |
|
产品技术规格
Appearance | Colorless to Yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...