Isobutyl isovalerate ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
Isobutyl Isovalerate, Isovaleric acid Isobutyl ester
I0197-25ML
I0197-500ML
TCIAI0197-25MLEA
0
CNY
TCIAI0197-25ML
TCIAI0197-500ML
Isobutyl isovalerate ≥98.0% (by GC)
Isobutyl isovalerate
分子式:
C₉H₁₈O₂ 沸点: 168°C 闪点: 57°C |
MDL编号:
MFCD00048349 CAS号: 589-59-3 EINECS: 209-653-8 UN号: 3272 ADR: 3,III Merck Index: 14,05146 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...