Ethyl methyl carbonate ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
Carbonic acid ethyl methyl ester
C1342-10ML
C1342-25ML
TCIAC1342-10MLEA
0
CNY
TCIAC1342-10ML
TCIAC1342-25ML
Ethyl methyl carbonate ≥98.0% (by GC)
Ethyl methyl carbonate
分子式:
C₄H₈O₃ 分子量: 104.1045 g/mol 沸点: 107°C |
MDL编号:
MFCD00191398 CAS号: 623-53-0 UN号: 3272 ADR: 3 III |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...