2-叔丁基蒽醌 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
2-tert-Butylanthraquinone
B0816-25G
B0816-500G
TCIAB0816-25GEA
0
CNY
TCIAB0816-25G
TCIAB0816-500G
2-叔丁基蒽醌 ≥98.0% (by GC)
2-叔丁基蒽醌
分子式:
C₁₈H₁₆O₂ 分子量: 264.32 g/mol |
MDL编号:
MFCD00001232 CAS号: 84-47-9 EINECS: 201-531-2 UN号: 3077 ADR: 9 III |
|
产品技术规格
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 101.0 to 104.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...