1,2,3,4-四氢-1-萘酚 ≥95.0% (通过GC)

供应商: TCI
Warning

T1111-25G T1111-5G
TCIAT1111-25GEA 0 CNY
TCIAT1111-25G TCIAT1111-5G
1,2,3,4-四氢-1-萘酚 ≥95.0% (通过GC)
1,2,3,4-四氢-1-萘酚
分子式: C₁₀H₁₂O
MDL编号: MFCD00001739
CAS号: 529-33-9
EINECS: 208-459-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8e19a9b044752487db9a990521917286cc8cc0f&vid=e8e19a9b044752487db9a990521917286cc8cc0f&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR