双季戊四醇 ≥80.0% (by GC)
供应商: TCI
Synonyms:
2,2'-(Oxybis(methylene))bis(2-(hydroxymethyl)propane-1,3-diol)
D1342-25G
D1342-500G
TCIAD1342-25GEA
0
CNY
TCIAD1342-25G
TCIAD1342-500G
双季戊四醇 ≥80.0% (by GC)
双季戊四醇
分子式:
C₁₀H₂₂O₇ |
MDL编号:
MFCD00004691 CAS号: 126-58-9 EINECS: 204-794-1 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 80.0 % |
Melting point | 217.0 to 222.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...