次氮基三乙酸二钠盐 ≥99.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: NTA, 氨三乙酸, N,N-双(羧甲基)甘氨酸

N0100-25G N0100-500G
TCIAN0100-25GEA 0 CNY
TCIAN0100-25G TCIAN0100-500G
次氮基三乙酸二钠盐 ≥99.0% (by titrimetric analysis)
次氮基三乙酸二钠盐
分子式: C₆H₇NNa₂O₆
分子量: 235.1 g/mol
MDL编号: MFCD00064230
CAS号: 15467-20-6
EINECS: 239-484-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc08779db4eb1fb3738ade8488dcbb4568e9f56e&vid=dc08779db4eb1fb3738ade8488dcbb4568e9f56e&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 99.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR