(R)-(+)-乳酰胺 ≥98.0% (by GC, total nitrogen)
供应商: TCI
Synonyms:
(R)-2-Hydroxypropanamide
L0167-25G
L0167-5G
TCIAL0167-25GEA
0
CNY
TCIAL0167-25G
TCIAL0167-5G
(R)-(+)-乳酰胺 ≥98.0% (by GC, total nitrogen)
(R)-(+)-乳酰胺
分子式:
C₃H₇NO₂ 分子量: 89.09 g/mol |
MDL编号:
MFCD00210310 CAS号: 598-81-2 |
|
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Optical purity (GC) | min. 99.0 ee% |
Purity (with Total Nitrogen) | min. 98.0 % |
Specific rotation [a]20/D | +18.0 to +23.0 deg (C=10, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...