1,4-二硫-2,5-二醇 ≥96.0% (by GC)

供应商: TCI

Synonyms: 二噻烷, 2,5-二羟基-1,4-二噻烷, 2,5-二羟基噻二烷

D1761-25G D1761-500G
TCIAD1761-25GEA 0 CNY
TCIAD1761-25G TCIAD1761-500G
1,4-二硫-2,5-二醇 ≥96.0% (by GC)
1,4-二硫-2,5-二醇
分子式: C₄H₈O₂S₂
分子量: 152.24 g/mol
存储条件: Refrigerator
MDL编号: MFCD00006659
CAS号: 40018-26-6
EINECS: 254-751-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e43dcb51379995973c72db4c66a8bd37e253447&vid=7e43dcb51379995973c72db4c66a8bd37e253447&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR