反式-2,4-癸二烯醛 ≥90.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
Decadienal, (2E,4E)-Deca-2,4-dienal, DDA
D1934-25ML
D1934-5ML
TCIAD1934-25MLEA
0
CNY
TCIAD1934-25ML
TCIAD1934-5ML
反式-2,4-癸二烯醛 ≥90.0% (by GC)
反式-2,4-癸二烯醛
分子式:
C₁₀H₁₆O 分子量: 152.24 g/mol 沸点: 116°C 闪点: 100°C |
MDL编号:
MFCD00007007 CAS号: 25152-84-5 EINECS: 246-668-9 |
|
产品技术规格
Appearance | Light yellow to Amber to Dark green clear liquid |
Purity (GC) | min. 90.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...