1-(Methylamino)anthraquinone ≥98.0% (by HPLC)

供应商: TCI
Warning

Synonyms: 1-(Methylamino)anthracene-9,10-dione

M0255-25G M0255-500G
TCIAM0255-25GEA 0 CNY
TCIAM0255-25G TCIAM0255-500G
1-(Methylamino)anthraquinone ≥98.0% (by HPLC)
1-(Methylamino)anthraquinone
分子式: C₁₅H₁₁NO₂
分子量: 237.26 g/mol
MDL编号: MFCD00001197
CAS号: 82-38-2
EINECS: 201-417-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0bd23909051c9c3575e1cc8003a27e97ee0e839&vid=d0bd23909051c9c3575e1cc8003a27e97ee0e839&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Orange to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR