水杨酸-2-乙基己基酯 ≥98.0% (通过GC)

供应商: TCI
Warning

S0387-25ML S0387-500ML
TCIAS0387-25MLEA 0 CNY
TCIAS0387-25ML TCIAS0387-500ML
水杨酸-2-乙基己基酯 ≥98.0% (通过GC)
水杨酸-2-乙基己基酯
分子式: C₁₅H₂₂O₃
沸点: 190°C
MDL编号: MFCD00053300
CAS号: 118-60-5
EINECS: 204-263-4
Merck Index: 14,06770
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=edd979e24e10d3c8b6061f03cbb12fd780805697&vid=edd979e24e10d3c8b6061f03cbb12fd780805697&prid=web&clid=SAPDAM

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR