2-氨基-5-三氟甲基-1,3,4-噻二唑 ≥98.0% (by HPLC)

供应商: TCI
Warning

Synonyms: 2-Amino-5-trifluoromethyl-1,3,4-thiadiazole

A2462-25G A2462-5G
TCIAA2462-25GEA 0 CNY
TCIAA2462-25G TCIAA2462-5G
2-氨基-5-三氟甲基-1,3,4-噻二唑 ≥98.0% (by HPLC)
2-氨基-5-三氟甲基-1,3,4-噻二唑
分子式: C₃H₂F₃N₃S
分子量: 169.13 g/mol
MDL编号: MFCD00003109
CAS号: 10444-89-0
EINECS: 233-930-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1ce212ac74edcd66b3ca03be8a1f217d229a638&vid=b1ce212ac74edcd66b3ca03be8a1f217d229a638&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 221.0 to 225.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR