N-Ethylrhodanine ≥98.0% (通过GC)

供应商: TCI
Warning

E0174-25G E0174-5G
TCIAE0174-25GEA 0 CNY
TCIAE0174-25G TCIAE0174-5G
N-Ethylrhodanine ≥98.0% (通过GC)
N-Ethylrhodanine
分子式: C₅H₇NOS₂
沸点: 128°C
存储条件: Refrigerator
MDL编号: MFCD00051339
CAS号: 7648-01-3
EINECS: 231-603-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=627e6d33a5ef1f4035fef4daa5f02673ed7f1508&vid=627e6d33a5ef1f4035fef4daa5f02673ed7f1508&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light yellow to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 98.0 %
Melting point 36.0 to 40.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR