偏苯三酸酐 ≥98.0% (by titrimetric analysis)
供应商: TCI
|
Danger
|
Synonyms:
偏苯三酸酐, 苯-1,2,4-三羧酸酐
C0959-25G
C0959-500G
TCIAC0959-25GEA
0
CNY
TCIAC0959-25G
TCIAC0959-500G
偏苯三酸酐 ≥98.0% (by titrimetric analysis)
偏苯三酸酐
分子式:
C₉H₄O₅ |
MDL编号:
MFCD00005925 CAS号: 552-30-7 EINECS: 209-008-0 Merck Index: 14,09703 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (Methanolysis Method) | min. 98.0 % |
Melting point | 166.0 to 170.0 deg-C |
Solubility in Acetone | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...