2,6-二甲基苯醌 ≥98.0% (by GC)

供应商: TCI

Synonyms: 2,6-Dimethylcyclohexa-2,5-diene-1,4-dione

D2234-1G D2234-25G D2234-5G
TCIAD2234-1GEA 0 CNY
TCIAD2234-1G TCIAD2234-25G TCIAD2234-5G
2,6-二甲基苯醌 ≥98.0% (by GC)
2,6-二甲基苯醌
分子式: C₈H₈O₂
分子量: 136.15 g/mol
MDL编号: MFCD00001605
CAS号: 527-61-7
EINECS: 208-420-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33a8696ca27b0646d4141b8f96009e1b6d33cac9&vid=33a8696ca27b0646d4141b8f96009e1b6d33cac9&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 71.0 to 74.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR