4-氨基-2,2,6,6-四甲基哌啶 ≥98.0% (by GC)
供应商: TCI
|
Danger
|
A1077-25ML
A1077-500ML
TCIAA1077-25MLEA
0
CNY
TCIAA1077-25ML
TCIAA1077-500ML
4-氨基-2,2,6,6-四甲基哌啶 ≥98.0% (by GC)
4-氨基-2,2,6,6-四甲基哌啶
分子式:
C₉H₂₀N₂ 分子量: 156.27 g/mol 沸点: 93°C 闪点: 81°C |
MDL编号:
MFCD00005984 CAS号: 36768-62-4 EINECS: 253-197-2 UN号: 2735 ADR: 8 III |
|
产品技术规格
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...