肼基甲酸苄酯 ≥98.0% (by titrimetric analysis)

供应商: TCI
Danger

Synonyms: 苄基肼基甲酸酯, 苄氧羰基肼

C1564-25G C1564-5G
TCIAC1564-25GEA 0 CNY
TCIAC1564-25G TCIAC1564-5G
肼基甲酸苄酯 ≥98.0% (by titrimetric analysis)
肼基甲酸苄酯
分子式: C₈H₁₀N₂O₂
分子量: 166.18 g/mol
MDL编号: MFCD00041890
CAS号: 5331-43-1
EINECS: 226-230-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f80f7f1f4dcda73073fd17ca913bc6c0a66dbe2&vid=6f80f7f1f4dcda73073fd17ca913bc6c0a66dbe2&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (Bromination Method) min. 98.0 %
Melting point 66.0 to 71.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR