肼基甲酸叔丁酯 ≥97.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: Boc-肼, 叔丁氧羰基肼

C0933-25G C0933-250G
TCIAC0933-25GEA 0 CNY
TCIAC0933-25G TCIAC0933-250G
肼基甲酸叔丁酯 ≥97.0% (by titrimetric analysis)
肼基甲酸叔丁酯
分子式: C₅H₁₂N₂O₂
分子量: 132.16 g/mol
沸点: 95°C
MDL编号: MFCD00007593
CAS号: 870-46-2
EINECS: 212-795-3
UN号: 1325
ADR: 4.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7bab6bf09e575a50568aaeec7c5eebb552f15863&vid=7bab6bf09e575a50568aaeec7c5eebb552f15863&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 39.0 to 43.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR