樟脑醌 ≥98.0% (by GC)

供应商: TCI

Synonyms: (±)-樟脑醌, 2,3-莰烷二酮

C0014-25G C0014-5G
TCIAC0014-25GEA 0 CNY
TCIAC0014-25G TCIAC0014-5G
樟脑醌 ≥98.0% (by GC)
樟脑醌
分子式: C₁₀H₁₄O₂
分子量: 166.22 g/mol
MDL编号: MFCD00064160
CAS号: 10373-78-1
EINECS: 233-814-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b29934255f62cd4f0e129274fcae1cd415a4dc7&vid=6b29934255f62cd4f0e129274fcae1cd415a4dc7&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light yellow to Yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 199.0 to 203.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR