1-氨基苯并三唑 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
ABT, 1-苯并三唑胺
A1464-1G
A1464-5G
TCIAA1464-1GEA
0
CNY
TCIAA1464-1G
TCIAA1464-5G
1-氨基苯并三唑 ≥98.0% (by GC)
1-氨基苯并三唑
分子式:
C₆H₆N₄ |
MDL编号:
MFCD00132902 CAS号: 1614-12-6 |
|
产品技术规格
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 81.0 to 85.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...