1-氨基-4-甲基哌嗪 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
4-Methylpiperazin-1-amine
A1192-25ML
A1192-500ML
TCIAA1192-25MLEA
0
CNY
TCIAA1192-25ML
TCIAA1192-500ML
1-氨基-4-甲基哌嗪 ≥98.0% (by GC, titration analysis)
1-氨基-4-甲基哌嗪
分子式:
C₅H₁₃N₃ 分子量: 115.18 g/mol 沸点: 175°C 闪点: 103°C |
MDL编号:
MFCD00006154 CAS号: 6928-85-4 EINECS: 230-053-7 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...