棕榈油酸 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 顺-9-十六烯酸

H0072-1G H0072-100MG
TCIAH0072-1GEA 0 CNY
TCIAH0072-1G TCIAH0072-100MG
棕榈油酸 ≥98.0% (by GC, titration analysis)
棕榈油酸
分子式: C₁₆H₃₀O₂
分子量: 254.41 g/mol
沸点: 175°C
溶点: 1°C
闪点: 62°C
存储条件: Refrigerator
MDL编号: MFCD00004437
CAS号: 373-49-9
EINECS: 206-765-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b234331a3a6bd412ec16643e90d2ec9b43e8494&vid=8b234331a3a6bd412ec16643e90d2ec9b43e8494&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR