棕榈油酸甲酯 ≥98.0% (by GC)

供应商: TCI

Synonyms: 顺式-9-十六碳烯酸甲酯, 棕榈油酸甲酯

P1958-1G P1958-100MG
TCIAP1958-1GEA 0 CNY
TCIAP1958-1G TCIAP1958-100MG
棕榈油酸甲酯 ≥98.0% (by GC)
棕榈油酸甲酯
分子式: C₁₇H₃₂O₂
分子量: 268.44 g/mol
沸点: 126°C
存储条件: Freezer
MDL编号: MFCD00042911
CAS号: 1120-25-8
EINECS: 214-303-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76275f2a36daf689277b56b2b9f92e086bb9feb6&vid=76275f2a36daf689277b56b2b9f92e086bb9feb6&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR