2-金刚烷酮 ≥98.0% (by GC)

供应商: TCI

Synonyms: Adamantan-2-one

A0719-100G A0719-25G A0719-5G
TCIAA0719-100GEA 0 CNY
TCIAA0719-100G TCIAA0719-25G TCIAA0719-5G
2-金刚烷酮 ≥98.0% (by GC)
2-金刚烷酮
分子式: C₁₀H₁₄O
分子量: 150.22 g/mol
沸点: 228
溶点: 43
存储条件: Ambient
MDL编号: MFCD00074737
CAS号: 700-58-3
EINECS: 211-847-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bac9e45c13df1a4753c4075ab5e09c9507cd6eae&vid=bac9e45c13df1a4753c4075ab5e09c9507cd6eae&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR