2,5-二甲基噻吩 ≥98.0% (by GC)

供应商: TCI
Warning

D1591-25ML D1591-5ML
TCIAD1591-25MLEA 0 CNY
TCIAD1591-25ML TCIAD1591-5ML
2,5-二甲基噻吩 ≥98.0% (by GC)
2,5-二甲基噻吩
分子式: C₆H₈S
分子量: 112.2 g/mol
沸点: 136°C
溶点: -63
闪点: 24°C
存储条件: Ambient
MDL编号: MFCD00005452
CAS号: 638-02-8
EINECS: 211-313-9
UN号: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e43b81bacb81482d1c22b211a5a09da10b85aa01&vid=e43b81bacb81482d1c22b211a5a09da10b85aa01&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR