3-辛基噻吩 ≥98.0% (by GC)

供应商: TCI

O0213-1G O0213-25G O0213-5G
TCIAO0213-1GEA 0 CNY
TCIAO0213-1G TCIAO0213-25G TCIAO0213-5G
3-辛基噻吩 ≥98.0% (by GC)
3-辛基噻吩
分子式: C₁₂H₂₀S
分子量: 196.36 g/mol
沸点: 106°C
存储条件: Refrigerator
MDL编号: MFCD00085281
CAS号: 65016-62-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59c9def5243500519ec8562655ae146b92ca32a6&vid=59c9def5243500519ec8562655ae146b92ca32a6&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR