3-氯噻吩 ≥97.0% (by GC)

供应商: TCI
Danger

C1192-25G C1192-5G
TCIAC1192-25GEA 0 CNY
TCIAC1192-25G TCIAC1192-5G
3-氯噻吩 ≥97.0% (by GC)
3-氯噻吩
分子式: C₄H₃ClS
分子量: 118.59 g/mol
沸点: 139°C
闪点: 37°C
MDL编号: MFCD00043887
CAS号: 17249-80-8
UN号: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb5aad5a542c4e019ab0d1067f38061395398baf&vid=fb5aad5a542c4e019ab0d1067f38061395398baf&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR