3-溴噻吩 ≥97.0% (by GC)

供应商: TCI
Danger

Synonyms: 3-噻吩基溴

B1067-25G B1067-250G
TCIAB1067-25GEA 0 CNY
TCIAB1067-25G TCIAB1067-250G
3-溴噻吩 ≥97.0% (by GC)
3-溴噻吩
分子式: C₄H₃BrS
沸点: 158°C
溶点: –10 °C
密度: 1,735 g/cm³ (20 °C)
闪点: 57°C
存储条件: Ambient
MDL编号: MFCD00005464
CAS号: 872-31-1
EINECS: 212-821-3
UN号: 1992
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7c837837c4b56ec6fdb7e2ee2ff8d0669288547&vid=b7c837837c4b56ec6fdb7e2ee2ff8d0669288547&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR