3,4-二溴噻吩 ≥98.0% (by GC)

供应商: TCI
Warning

D1676-25G D1676-5G
TCIAD1676-25GEA 0 CNY
TCIAD1676-25G TCIAD1676-5G
3,4-二溴噻吩 ≥98.0% (by GC)
3,4-二溴噻吩
分子式: C₄H₂Br₂S
分子量: 241.93 g/mol
沸点: 100°C
溶点: 5°C
存储条件: Ambient
MDL编号: MFCD00005465
CAS号: 3141-26-2
EINECS: 221-546-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a71ffe24d6eb0747b02a29e2fb8852895ba6d4d1&vid=a71ffe24d6eb0747b02a29e2fb8852895ba6d4d1&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR