2-溴噻吩 ≥98.0% (by GC)
供应商: TCI
|
Danger
|
Synonyms:
2-噻吩基溴
B0941-25G
B0941-250G
TCIAB0941-25GEA
0
CNY
TCIAB0941-25G
TCIAB0941-250G
2-溴噻吩 ≥98.0% (by GC)
2-溴噻吩
分子式:
C₄H₃BrS 分子量: 163.04 g/mol 沸点: 154°C 溶点: -10 闪点: 60°C 存储条件: Refrigerator |
MDL编号:
MFCD00005417 CAS号: 1003-09-4 EINECS: 213-699-4 UN号: 2929 ADR: 6.1 II |
|
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...