2,3,5-可力丁 ≥98.0% (by GC)
供应商: TCI
Warning
Synonyms: 2,3,5-三甲基吡啶
T1722-25G T1722-5G
TCIAT1722-25GEA 0 CNY
TCIAT1722-25G TCIAT1722-5G
2,3,5-可力丁 ≥98.0% (by GC)
2,3,5-可力丁
分子式: C₈H₁₁N
分子量: 121.18 g/mol
沸点: 65°C
闪点: 74°C
MDL编号: MFCD00126840
CAS号: 695-98-7
EINECS: 211-786-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb45952f6181abd7df14944e65e2edc9ace13774&vid=fb45952f6181abd7df14944e65e2edc9ace13774&prid=web&clid=SAPDAM
现在订购
产品技术规格
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %