2-氯-4-甲基吡啶 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
2-氯-4-皮考啉
C1992-25G
C1992-5G
TCIAC1992-25GEA
0
CNY
TCIAC1992-25G
TCIAC1992-5G
2-氯-4-甲基吡啶 ≥98.0% (by GC)
2-氯-4-甲基吡啶
分子式:
C₆H₆ClN 分子量: 127.57 g/mol 沸点: 99°C 溶点: 115 |
MDL编号:
MFCD00023418 CAS号: 3678-62-4 EINECS: 222-951-2 |
|
产品技术规格
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...