3,4-二氨基吡啶 ≥99.0% (by GC, titration analysis)
供应商: TCI
|
Danger
|
Synonyms:
3,4-Pyridinediamine, Amifampridine, 3,4-DAP, Pyridine-3,4-diyldiamine, Pyridine-3,4-diamine
D1149-1G
D1149-5G
TCIAD1149-1GEA
0
CNY
TCIAD1149-1G
TCIAD1149-5G
3,4-二氨基吡啶 ≥99.0% (by GC, titration analysis)
3,4-二氨基吡啶
分子式:
C₅H₇N₃ 分子量: 109.13 g/mol 沸点: 262 闪点: 240 存储条件: Ambient |
MDL编号:
MFCD00006401 CAS号: 54-96-6 EINECS: 200-220-9 UN号: 2811 ADR: 6.1 II Merck Index: 14,02986 |
|
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 99.0 % |
Purity (Nonaqueous Titration) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...