五氟吡啶 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 2,3,4,5,6-Perfluoropyridine, 2,3,4,5,6-Pentafluoropyridine

P0926-25G P0926-5G
TCIAP0926-25GEA 0 CNY
TCIAP0926-25G TCIAP0926-5G
五氟吡啶 ≥98.0% (by GC)
五氟吡啶
分子式: C₅F₅N
沸点: 83°C
溶点: –41 °C
密度: 1,54 g/cm³ (20 °C)
闪点: 25°C
存储条件: Ambient
MDL编号: MFCD00006225
CAS号: 700-16-3
EINECS: 211-839-9
UN号: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=133c313fe9c42ef327f060eb57fbe6643378784d&vid=133c313fe9c42ef327f060eb57fbe6643378784d&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR