5-溴-2-氯吡啶 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
B2329-25G
B2329-5G
TCIAB2329-25GEA
0
CNY
TCIAB2329-25G
TCIAB2329-5G
5-溴-2-氯吡啶 ≥98.0% (by GC)
5-溴-2-氯吡啶
分子式:
C₅H₃BrClN 分子量: 192.44 g/mol 沸点: 205 溶点: 71 |
MDL编号:
MFCD01318951 CAS号: 53939-30-3 |
|
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 67.0 to 75.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...