2-Bromo-6-chloropyridine ≥98.0% (by GC)

供应商: TCI
Warning

B3609-1G B3609-5G
TCIAB3609-1GEA 0 CNY
TCIAB3609-1G TCIAB3609-5G
2-Bromo-6-chloropyridine ≥98.0% (by GC)
2-Bromo-6-chloropyridine
分子式: C₅H₃BrClN
分子量: 192.44 g/mol
MDL编号: MFCD00181262
CAS号: 5140-72-7
EINECS: 225-904-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de4e1462f557082b0dedaeb02767f425bfc71ea9&vid=de4e1462f557082b0dedaeb02767f425bfc71ea9&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 88.0 to 92.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR